Learn More
Sino Biological Recombinant Human ICOS Protein (His & hFc Tag), HPLC-verified
Supplier:  Sino Biological 10344H03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human ICOS (NP_036224.1) (Met 1-Phe 141) was fused to the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4671