Learn More
Sino Biological Recombinant Human ICOS Protein (rFc Tag)
Supplier:  Sino Biological 10344H31H100
Encompass_Preferred
A DNA sequence encoding the human ICOS (NP_036224.1) (Met1-Phe141) was expressed with the Fc region of rabbit IgG at the C-terminus.
Catalog No. 50-161-4673