Learn More
Sino Biological Anti-Granzyme B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10345RP03100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Granzyme B (rhGranzyme B; Catalog#10345-H08H; NP_004122.1; Met 1-Tyr 247). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-4773