Learn More
Sino Biological Recombinant Human ICAM-1/CD54 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10346H08H50
Encompass_Preferred
A DNA sequence encoding the human ICAM1 (NP_000192.2) extracellular domain (Met 1-Glu 480) was fused with the a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4680