Learn More
Sino Biological Anti-ICAM-1/CD54 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10346MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ICAM1 / CD54 (rh ICAM1 / CD54; Catalog#10346-H08H; Met 1-Glu 480; NP_000192.2). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.ved.
Catalog No. 50-172-4792