Learn More
Sino Biological Anti-ICAM-1/CD54 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10346MM01F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ICAM-1 / CD54 (rh ICAM-1 / CD54; Catalog#10346-H08H; NP_000192.2; Met 1-Glu 480) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-4800