Learn More
Sino Biological Anti-ICAM-1/CD54 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10346RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ICAM1 / CD54 extracellular domain (rh ICAM1; Catalog#10346-H08H; NP_000192.2; Met 1-Glu 480). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-4808