Learn More
Sino Biological Recombinant Human IFN-alpha B2/IFNA8 Protein
Supplier:  Sino Biological 10347HNCH100
Encompass_Preferred
A DNA sequence encoding the human IFNA8 (NP_002161.2)(Cys24-Glu189) was expressed and purified with two additional amino acids (Gly & Pro ) at the N-terminus.
Catalog No. 50-161-4685