Learn More
Sino Biological Recombinant Anti-IFN-alpha B2/IFNA8 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10347R003100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Interferon alpha-B / IFNA8 (rh Interferon alpha-B / IFNA8; Catalog#10347-HNCH; NP_002161.2; Cys24-Glu189).
Catalog No. 50-172-4829