Learn More
Sino Biological Anti-IFN-alpha B2/IFNA8 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10347T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Interferon alpha-B / IFNA8 (rh Interferon alpha-B / IFNA8; Catalog#10347-H08H; NP_002161.2; Met1-Glu189). Interferon alpha-B / IFNA8 specific IgG was purified by Human Interferon alpha-B / IFNA8 affinity chromatography.
Catalog No. 50-172-4834