Learn More
Sino Biological Anti-Granzyme H Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10348RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Granzyme H / GZMH (rh GZMH; Catalog#10348-H08H; NP_219491.1; Met1-Leu246). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-4843