Learn More
Sino Biological Recombinant Human IL-13 R alpha 2/IL13RA2 Protein (His & hFc Tag)
Supplier:  Sino Biological 10350H03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human IL13Rα2 (NP_000631.1) (Met 1-Leu 342) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4696