Learn More
Sino Biological Recombinant Human IL-13 R alpha 2/IL13RA2 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10350H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human IL13Rα2 (NP_000631.1) (Met1-Leu342) was expressed with the a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4698