Learn More
Sino Biological Recombinant Anti-IL-13 R alpha 2/IL13RA2 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 10350R018A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL13RA2 / CD213A2 (rh IL13RA2 / CD213A2; Catalog#10350-H08H; NP_000631.1; Met1-Leu342) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-172-4854