Learn More
Sino Biological Anti-IL-13 R alpha 2/IL13RA2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10350T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL13RA2 / CD213A2 (rh IL13RA2 / CD213A2; Catalog#10350-H08H; NP_000631.1; Met1-Leu342). IL13RA2 / CD213A2 specific IgG was purified by Human IL13RA2 / CD213A2 affinity chromatography.
Catalog No. 50-172-4863