Learn More
Sino Biological Anti-Hsp27/HSPB1 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10351MM09F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human HSP27 / HSPB1 (rh HSP27 / HSPB1; Catalog#10351-H08E; NP_001532.1; Met1-Lys205) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-4874