Learn More
Sino Biological Recombinant Anti-Hsp27/HSPB1 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10351R155F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human HSP27 (rh HSP27; Catalog#10351-H08E; NP_001532.1; Met1-Lys205) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-4881