Learn More
Sino Biological Anti-Interferon omega-1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10353MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IFN omega / IFNW1 (rh IFN omega / IFNW1; Catalog#10353-H08H; P05000; Met1-Ser195). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-4912