Learn More
Sino Biological Anti-Interferon omega-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10353T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IFN omega (rh IFN omega; Catalog#10353-H08H; P05000; Met1-Ser195). IFN omega specific IgG was purified by Human IFN omega affinity chromatography.
Catalog No. 50-172-4918