Learn More
Sino Biological Recombinant Human RBP4 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10354H08H100
Encompass_Preferred
A DNA sequence encoding the pro form of human RBP4 (NP_006735.2) (Met 1-Leu 201) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4708