Learn More
Sino Biological Anti-RBP4 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10354MM15P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human RBP4 (rh RBP4; Catalog#10354-H08H; NP_006735.2; Met 1-Leu 201) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-172-4938