Learn More
Sino Biological Anti-RBP4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10354RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human RBP4 / PRBP (rhRBP4; Catalog#10354-H08H; Met 1-Leu 201; NP_006735.2). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-4939