Learn More
Sino Biological Recombinant Human Survivin/BIRC5 Protein
Supplier:  Sino Biological 10356HNCE100
Encompass_Preferred
A DNA sequence encoding the human BIRC5 (NP_001159.2) (Met1-Asp142) was expressed and purified with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-4712