Learn More
Sino Biological Anti-IL-18BP Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10357RP04100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL18BP N-terminal domain (rhIL18BP; Catalog#10357-H08H; NP_001034748.1; Met1-Gly194). IL18BP specific IgG was purified by human IL18BP affinity chromatography.
Catalog No. 50-172-4971