Learn More
Sino Biological Recombinant Human TGF-beta RII/TGFBR2 Protein (ECD, His Tag)
Supplier:  Sino Biological 10358H08B100
Encompass_Preferred
A DNA sequence encoding the human TGFBR2 (NP_003233.4) (Met1-Gln166) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4721