Learn More
Sino Biological Recombinant Human Interferon alpha/beta receptor 2 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 10359H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human IFNAR2 isoform a (NP_997468.1) (Met 1-Lys 243) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4723