Learn More
Sino Biological Recombinant Human Interferon alpha/beta receptor 2 Protein (His Tag), Amine-Labeled Biotinylated, HPLC-verified
Supplier:  Sino Biological 10359H08HB20
Encompass_Preferred
A DNA sequence encoding the human IFNAR2 (NP_997468.1) (Met1-Lys243) was expressed with a polyhistidine tag at the C-terminus.The expressed protein was biotinylated in vitro.
Catalog No. 50-161-4727