Learn More
Sino Biological Anti-Interferon alpha/beta receptor 2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10359MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IFNAR2 (rh IFNAR2; Catalog#10359-H08H; NP_997468.1; Met1-Lys243). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-4983