Learn More
Sino Biological Anti-C19orf47 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 103597T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human C19orf47, and purified by antigen affinity chromatography.
Catalog No. 50-172-4980