Learn More
Sino Biological Recombinant Human IL-15 Protein (His Tag)
Supplier:  Sino Biological 10360H07E100
Encompass_Preferred
A DNA sequence encoding the human IL15 (NP_000576.1) (Asn49-Ser162) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-4729