Learn More
Sino Biological Anti-IL-15 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10360RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL-15 / IL15 / Interleukin 15 (rh IL-15 / IL15 / Interleukin 15; Catalog#10360-H07E; NP_000576.1; Asn49-Ser162). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5012