Learn More
Sino Biological Anti-ECM1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10362RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ECM1 (rhECM1; Catalog#10362-H08H; Met 1-Glu 540; NP_004416.1). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5033