Learn More
Sino Biological Recombinant Human, Rhesus ERBB4/Her4 Protein (hFc Tag)
Supplier:  Sino Biological 10363H02H100
Encompass_Preferred
A DNA sequence encoding the human / rhesus ERBB4 (NP_005226.1) (Met1-Arg649) was expressed, fused with the Fc region of human IgG1 at the C-terminus. Human and Rhesus ERBB4 sequences are identical.
Catalog No. 50-161-4733