Learn More
Sino Biological Anti-MOG Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10364MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MOG (rh MOG; Catalog#10364-H08E; NP_996532.2; Gly30-Tyr149). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-5042