Learn More
Sino Biological Recombinant Human IL-33 Protein, HPLC-verified
Supplier:  Sino Biological 10368HNAE5
Encompass_Preferred
A DNA sequence encoding the C-terminal segment of human IL33 (NP_254274.1) (Ser 112-Thr 270) was expressed with an initial Met at the N-terminus.
Catalog No. 50-161-4745