Learn More
Sino Biological Anti-IL-33 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10368MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-33 (rh IL-33; Catalog#10368-HNAE; NP_254274.1; Ser 112-Thr 270). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-5053