Learn More
Sino Biological Anti-IL-33 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10368RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL-33 (rh IL-33; Catalog#10368-HNAE; Ser 112-Thr 270; NP_254274.1). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5083