Learn More
Sino Biological Recombinant Human IL-13 Protein, HPLC-verified
Supplier:  Sino Biological 10369HNAC5
Encompass_Preferred
A DNA sequence encoding the human IL13 (AAK53823.1) (Met 1-Asn 132) was expressed and purified.
Catalog No. 50-161-4751