Learn More
Sino Biological Anti-IL-13 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10369RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL13 / ALRH (rh IL13 / ALRH; Catalog#10369-H01H; AAK53823.1; Gly21-Asn132). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-5108