Learn More
Sino Biological Recombinant Human Sonic Hedgehog/SHH Protein (aa 198-462, His Tag)
Supplier:  Sino Biological 10372H08H100
Encompass_Preferred
A DNA sequence encoding the amino acid sequence (Cys 198-Ser 462) of human SHH (Q15465), that is Sonic hedgehog protein C-product, was fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-161-4755