Learn More
Sino Biological Recombinant Human Sonic Hedgehog/SHH Protein (aa 1-197, His Tag), HPLC-verified
Supplier:  Sino Biological 10372H08H120
Encompass_Preferred
A DNA sequence encoding the amino acid sequence (Met 1-Gly 197) of human SHH (Q15465), that is Sonic hedgehog protein N-product, was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4758