Learn More
Sino Biological Recombinant Human CD32A/FCGR2A Protein (R167, His Tag), HPLC-verified
Supplier:  Sino Biological 10374H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Ile218) of human FCGR2A (P12318-1, with Natural variant R167) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4771