Learn More
Sino Biological CD32/FCGR2/Fc gamma RII Antibody, Mouse MAb
Supplier:  Sino Biological 10374MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD32a extracellular domain (rh CD32a; Catalog#10374-H08H; P12318-1; Met1-Ile218,167R). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-5123