Learn More
Sino Biological CD32/FCGR2/Fc gamma RII Antibody (APC), Mouse MAb
Supplier:  Sino Biological 10374MM01A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD32a/FCGR2A/Fc gamma RIIA (rh CD32a/FCGR2A/Fc gamma RIIA; Catalog#10374-H08H; P12318-1; Met1-Ile218,167R) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-172-5127