Learn More
Sino Biological Anti-CD32A/FCGR2A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10374RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD32a / Fc gamma RIIA (rh CD32a / Fc gamma RIIA; Catalog#10374-H08H; P12318-1; Met1-Ile218,167R). CD32a / Fc gamma RIIA specific IgG was purified by Human CD32a / Fc gamma RIIA
Catalog No. 50-172-5151