Learn More
Sino Biological Recombinant Human PD-1 Protein (ECD, mFc Tag), HPLC-verified
Supplier:  Sino Biological 10377H05H50
Encompass_Preferred
A DNA sequence encoding the human PDCD1 (NP_005009.2) (Met1-Gln167) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-4789