Learn More
Sino Biological PD1 / PDCD1 / CD279 Neutralizing Antibody
Supplier:  Sino Biological 10377HF06200
Encompass_Preferred
This antibody was a humanized antibody obtained from a mouse immunized with the recombinant Human PD1 / PDCD1 Protein (Catalog#10377-H08H; NP_005009.2; Met 1-Gln 167) and was produced using recombinant antibody technology.
Catalog No. 50-172-5160