Learn More
Sino Biological Anti-PD-1 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10377MA04P25
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells, and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-172-5175