Learn More
Sino Biological Recombinant Human TPO/Thrombopoietin Protein (hFc Tag)
Supplier:  Sino Biological 10381H02H100
Encompass_Preferred
A DNA sequence encoding the human THPO (NP_000451.1) (Met1-Leu192) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4794