Learn More
Sino Biological Recombinant Human TPO/Thrombopoietin Protein (His Tag)
Supplier:  Sino Biological 10381H08C100
Encompass_Preferred
A DNA sequence encoding the human THPO (NP_000451.1) precursor (Met 1-Gly 353) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4799